2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-cellobiosyl bromide ≥98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(((2R,3R,4S,5R,6R)-4,5-diacetoxy-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate, Acetobromocellobiose

A785103-1G A785103-250MG A785103-100MG A785103-5G
77627-564EA 81.37 USD
77627-564 77627-566 77878-632 77878-633
2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-cellobiosyl bromide ≥98%
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide

Precaución: for Research Use Only

Formula: C₂₆H₃₅BrO₁₇
MDL Number: MFCD00069839
CAS Number: 14227-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR