3-Bromo-1-methyl-1H-pyrazole-5-carboxylic acid ≥95%
Supplier: Ambeed
|
Warning
|
A784919-5G
A784919-100MG
A784919-1G
A784919-250MG
77893-546EA
1029
USD
77893-546
77893-543
77893-544
77893-545
3-Bromo-1-methyl-1H-pyrazole-5-carboxylic acid ≥95%
3-Bromo-1-methyl-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₅BrN₂O₂ |
MDL Number:
MFCD22395741 CAS Number: 173841-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...