5-(6-Hydroxybenzofuran-2-yl)-2-(3-methylbut-2-en-1-yl)benzene-1,3-diol ≥98%

Supplier: Ambeed
Warning

Synonyms: Moracin C

A784508-1MG A784508-10MG A784508-5MG
77614-402EA 154.5 USD
77614-402 77686-800 77686-801
5-(6-Hydroxybenzofuran-2-yl)-2-(3-methylbut-2-en-1-yl)benzene-1,3-diol ≥98%
5-(6-Hydroxybenzofuran-2-yl)-2-(3-methylbut-2-en-1-yl)benzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₉H₁₈O₄
MW: 310.34 g/mol
MDL Number: MFCD20274766
CAS Number: 69120-06-5
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR