N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide maleate 98+%

Supplier: Ambeed
Danger

Synonyms: Betrixaban maleate

A784396-100MG A784396-250MG
77618-162EA 44.29 USD
77618-162 77618-164
N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide maleate 98+%
N-(5-Chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide maleate

Precaución: for Research Use Only

Formula: C₂₇H₂₆ClN₅O₇
MW: 567.9777 g/mol
MDL Number: MFCD31729381
CAS Number: 936539-80-9
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR