4-Cyanophenyl-4-ethylbenzoate ≥97%

Supplier: Ambeed
Warning

A784226-5G A784226-25G
77674-140EA 15 USD
77674-140 77873-245
4-Cyanophenyl-4-ethylbenzoate ≥97%
4-Cyanophenyl 4-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₂
MDL Number: MFCD00429470
CAS Number: 56131-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR