Potassium trifluoro(1-methylindol-2-yl)borate ≥95%

Supplier: Ambeed
Warning

A783960-5G A783960-1G A783960-250MG
77784-020EA 823 USD
77784-020 77784-018 77784-019
Potassium trifluoro(1-methylindol-2-yl)borate ≥95%
Potassium trifluoro(1-methylindol-2-yl)borate

Precaución: for Research Use Only

Formula: C₉H₈BF₃KN
MDL Number: MFCD29764431
CAS Number: 1428884-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR