1-Fluoro-2-iodo-4-methoxybenzene ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-iodoanisole

A783737-250MG A783737-5G A783737-100MG A783737-1G
77886-754EA 65 USD
77886-754 77886-755 77886-752 77886-753
1-Fluoro-2-iodo-4-methoxybenzene ≥97%
1-Fluoro-2-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₆FIO
MDL Number: MFCD16295276
CAS Number: 1028263-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR