tert-Butyl 5,8,11,14-tetraoxa-2-azaheptadecan-17-oate ≥97%

Supplier: Ambeed
Warning

Synonyms: Methylamino-PEG4-Boc

A783717-100MG A783717-250MG A783717-1G
77618-154EA 85.49 USD
77618-154 77618-156 77749-549
tert-Butyl 5,8,11,14-tetraoxa-2-azaheptadecan-17-oate ≥97%
tert-Butyl 5,8,11,14-tetraoxa-2-azaheptadecan-17-oate

Precaución: for Research Use Only

Formula: C₁₆H₃₃NO₆
MDL Number: MFCD28155210
CAS Number: 1621616-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR