Methyl 3-bromobicyclo[1.1.1]pentane-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A782960-250MG A782960-100MG
77573-520EA 225.57 USD
77573-520 77749-547
Methyl 3-bromobicyclo[1.1.1]pentane-1-carboxylate ≥98%
Methyl 3-bromobicyclo[1.1.1]pentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉BrO₂
MW: 205.05 g/mol
MDL Number: MFCD31620784
CAS Number: 83249-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR