2,2-Bis(2-hydroxy-5-biphenylyl)propane ≥98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Isopropylidenebis(2-phenylphenol), 5,5'-Isopropylidenebis(2-hydroxybiphenyl), 5,5''-(Propane-2,2-diyl)bis(([1,1'-biphenyl]-2-ol)), 2,2-Bis(3-phenyl-4-hydroxyphenyl)propane

A782735-1G A782735-250MG A782735-5G
77603-008EA 38.95 USD
77603-008 77882-692 77882-693
2,2-Bis(2-hydroxy-5-biphenylyl)propane ≥98%
2,2-Bis(2-hydroxy-5-biphenylyl)propane

Precaución: for Research Use Only

Formula: C₂₇H₂₄O₂
MDL Number: MFCD00272620
CAS Number: 24038-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR