Cycloheptylacetic acid ≥90%

Supplier: Ambeed
Danger

A782680-1G A782680-250MG
77603-006EA 66.63 USD
77603-006 77627-560
Cycloheptylacetic acid ≥90%
Cycloheptylacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₆O₂
MDL Number: MFCD01075701
CAS Number: 4401-20-1
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR