Bis-PEG1-NHS ester ≥98%

Supplier: Ambeed
Warning

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) 3,3'-oxydipropanoate

A782582-1G A782582-250MG A782582-5G
77686-796EA 263 USD
77686-796 77686-797 77686-798
Bis-PEG1-NHS ester ≥98%
Bis-PEG1-NHS ester

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₉
MW: 356.285 g/mol
MDL Number: MFCD20226397
CAS Number: 65869-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR