6-Methyl-1H-indazole-5-boronic acid ≥98%

Supplier: Ambeed
Warning

A782274-1G A782274-250MG A782274-5G
77686-795EA 112 USD
77686-795 77901-771 77901-226
6-Methyl-1H-indazole-5-boronic acid ≥98%
6-Methyl-1H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD12028645
CAS Number: 1310405-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR