N-(4-Aminobutyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Thalidomide-O-amido-C4-NH2
A781949-250MG
A781949-100MG
A781949-50MG
77618-150EA
402.73
USD
77618-150
77749-544
77749-545
N-(4-Aminobutyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide ≥99%
N-(4-Aminobutyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide
Precaución: for Research Use Only
Formula:
C₁₉H₂₂N₄O₆ MW: 402.4 g/mol |
MDL Number:
MFCD31630818 CAS Number: 1799711-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...