2-(Aminomethyl)-6-(trifluoromethyl)pyridine ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(6-(Trifluoromethyl)pyridin-2-yl)methanamine
A781770-1G
A781770-250MG
77627-552EA
515
USD
77627-552
77627-554
2-(Aminomethyl)-6-(trifluoromethyl)pyridine ≥98%
2-(Aminomethyl)-6-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₇H₇F₃N₂ MW: 176.139 g/mol |
MDL Number:
MFCD10696232 CAS Number: 916304-19-3 UN: 2735 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...