SU-5402 ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-[3-(2-Carboxyethyl)-4-methylpyrrol-2-methylidenyl]-2-indolinone, SU 5402, 3-(4-Methyl-2-((2-oxoindolin-3-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid

A781751-10MG A781751-1MG A781751-5MG
77614-386EA 98.88 USD
77614-386 77614-388 77614-390
SU-5402 ≥98%
SU-5402

Precaución: for Research Use Only

MW: 296.3 g/mol
MDL Number: MFCD08235144
CAS Number: 215543-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR