Tris(trimethylsilyl)silane ≥95% stabilised

Supplier: Ambeed
Danger

Synonyms: 1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)trisilane, TTMSS

Stabilization: Stabilised with Tetrabromobisphenol A (TBBPA) 0.05%

A781447-5G A781447-25G
77573-512EA 112 USD
77573-512 77873-242
Tris(trimethylsilyl)silane ≥95% stabilised
Tris(trimethylsilyl)silane

Precaución: for Research Use Only

Formula: C₉H₂₈Si₄
MDL Number: MFCD00077893
CAS Number: 1873-77-4
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR