4-(4-((6-(3,5-Dihydroxyphenoxy)-4,7,9-trihydroxydibenzo[b,e][1,4]dioxin-2-yl)oxy)-3,5-dihydroxyphenoxy)dibenzo[b,e][1,4]dioxine-1,3,6,8-tetraol ≥98%

Supplier: Ambeed
Warning

Synonyms: Dieckol

A780751-10MG A780751-5MG A780751-1MG A780751-2MG
77749-540EA 475 USD
77749-540 77749-543 77749-541 77749-542
4-(4-((6-(3,5-Dihydroxyphenoxy)-4,7,9-trihydroxydibenzo[b,e][1,4]dioxin-2-yl)oxy)-3,5-dihydroxyphenoxy)dibenzo[b,e][1,4]dioxine-1,3,6,8-tetraol ≥98%
4-(4-((6-(3,5-Dihydroxyphenoxy)-4,7,9-trihydroxydibenzo[b,e][1,4]dioxin-2-yl)oxy)-3,5-dihydroxyphenoxy)dibenzo[b,e][1,4]dioxine-1,3,6,8-tetraol

Precaución: for Research Use Only

Formula: C₃₆H₂₂O₁₈
MW: 742.55 g/mol
MDL Number: MFCD20232943
CAS Number: 88095-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR