1-(3-Iodophenyl)-4-(3-nitrophenyl)-1H-1,2,3-triazole ≥98%

Supplier: Ambeed
Warning

Synonyms: P62-mediated mitophagy inducer

A780511-10MG A780511-1MG A780511-5MG
77618-144EA 57.68 USD
77618-144 77618-146 77618-148
1-(3-Iodophenyl)-4-(3-nitrophenyl)-1H-1,2,3-triazole ≥98%
1-(3-Iodophenyl)-4-(3-nitrophenyl)-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₁₄H₉IN₄O₂
MW: 392.15 g/mol
MDL Number: MFCD31620757
CAS Number: 1809031-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR