CBR-5884 ≥99%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-(furan-2-carboxamido)-3-methyl-4-thiocyanatothiophene-2-carboxylate

A780427-5MG A780427-1MG A780427-10MG
77618-142EA 35.02 USD
77618-142 77618-140 77747-526
CBR-5884 ≥99%
CBR-5884

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₄S₂
MDL Number: MFCD04451442
CAS Number: 681159-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR