Coumarin 102 ≥97%

Supplier: Ambeed
Warning

Synonyms: 2,3,6,7-Tetrahydro-9-methyl-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one, 9-Methyl-2,3,6,7-tetrahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinolin-11(5H)-one, Coumarin 480

A779463-1G A779463-250MG
77573-510EA 123 USD
77573-510 77618-136
Coumarin 102 ≥97%
Coumarin 102

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₂
MDL Number: MFCD00041844
CAS Number: 41267-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR