4-Isopropoxy-3-methylphenylboronic acid pinacol ester ≥97%

Supplier: Ambeed
Warning

A779417-5G A779417-250MG A779417-1G
77749-539EA 480 USD
77749-539 77749-538 77749-537
4-Isopropoxy-3-methylphenylboronic acid pinacol ester ≥97%
4-Isopropoxy-3-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₃
MDL Number: MFCD25953916
CAS Number: 2121514-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR