(S)-3-(Boc-amino)-3-(4-methylphenyl)propionic acid ≥95%

Supplier: Ambeed
Warning

A778812-1G A778812-5G
77882-682EA 152 USD
77882-682 77882-683
(S)-3-(Boc-amino)-3-(4-methylphenyl)propionic acid ≥95%
(S)-3-(Boc-amino)-3-(4-methylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD03427897
CAS Number: 479064-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR