4-(Pyridin-2-ylmethyl)aniline dihydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: [4-(Pyridin-2-ylmethyl)phenyl]amine dihydrochloride

A777732-100MG A777732-250MG
77882-680EA 22 USD
77882-680 77882-681
4-(Pyridin-2-ylmethyl)aniline dihydrochloride ≥97%
4-(Pyridin-2-ylmethyl)aniline dihydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂N₂
MW: 257.16 g/mol
MDL Number: MFCD08070700
CAS Number: 96616-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR