tert-Butyl 9-hydroxy-3-azaspiro[5.5]undecane-3-carboxylate ≥98%

Supplier: Ambeed
Warning

A777612-250MG A777612-5G
77614-382EA 50.47 USD
77614-382 77674-130
tert-Butyl 9-hydroxy-3-azaspiro[5.5]undecane-3-carboxylate ≥98%
tert-Butyl 9-hydroxy-3-azaspiro[5.5]undecane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₃
MW: 269.38 g/mol
MDL Number: MFCD12755293
CAS Number: 918644-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR