(4-(Piperazin-1-yl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A777314-250MG A777314-100MG A777314-1G A777314-5G
77602-998EA 104.55 USD
77602-998 77878-623 77878-624 77878-625
(4-(Piperazin-1-yl)phenyl)boronic acid ≥98%
(4-(Piperazin-1-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BN₂O₂
MW: 206.05 g/mol
MDL Number: MFCD09608131
CAS Number: 513246-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR