2-Bromo-3-iodo-4-methylpyridine ≥95%

Supplier: Ambeed
Warning

A777034-5G A777034-100MG A777034-250MG
77674-128EA 3557 USD
77674-128 77884-715 77884-716
2-Bromo-3-iodo-4-methylpyridine ≥95%
2-Bromo-3-iodo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MDL Number: MFCD04112506
CAS Number: 884494-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR