Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-2-carboxylate 95+%
Supplier: Ambeed
|
Warning
|
A776749-100MG
A776749-250MG
A776749-5G
A776749-1G
77670-084EA
90
USD
77670-084
77873-234
77873-235
77873-233
Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-2-carboxylate 95+%
Methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]thiophene-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₆H₁₉BO₄S |
MDL Number:
MFCD05865112 CAS Number: 690632-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...