Bis(2-diphenylphosphinoethyl)phenylphosphine ≥98%

Supplier: Ambeed
Danger

Synonyms: ((Phenylphosphinediyl)bis(ethane-2,1-diyl))bis(diphenylphosphine), TRIPHOS

A776375-250MG A776375-1G
77602-996EA 35.88 USD
77602-996 77627-542
Bis(2-diphenylphosphinoethyl)phenylphosphine ≥98%
Bis(2-diphenylphosphinoethyl)phenylphosphine

Precaución: for Research Use Only

Formula: C₃₄H₃₃P₃
MW: 534.559 g/mol
MDL Number: MFCD00003048
CAS Number: 23582-02-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR