6-Bromo-2-phenylquinoline ≥95%

Supplier: Ambeed
Warning

A774558-1G A774558-5G
77882-676EA 51 USD
77882-676 77882-677
6-Bromo-2-phenylquinoline ≥95%
6-Bromo-2-phenylquinoline

Precaución: for Research Use Only

Formula: C₁₅H₁₀BrN
MDL Number: MFCD17392928
CAS Number: 3894-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR