1-Bromo-4-iodo-2,5-dimethylbenzene ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,5-dimethyl-4-iodobenzene

A774515-1G A774515-5G
77595-226EA 33.83 USD
77595-226 77684-713
1-Bromo-4-iodo-2,5-dimethylbenzene ≥97%
1-Bromo-4-iodo-2,5-dimethylbenzene

Precaución: For research use only.

Formula: C₈H₈BrI
MW: 310.95 g/mol
MDL Number: MFCD07780644
CAS Number: 699119-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR