Ethyl 4-bromo-1,3-thiazole-2-carboxylate ≥95%

Supplier: Ambeed
Warning

A774293-1G A774293-5G A774293-25G
77614-372EA 38.11 USD
77614-372 77614-374 77886-741
Ethyl 4-bromo-1,3-thiazole-2-carboxylate ≥95%
Ethyl 4-bromo-1,3-thiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MDL Number: MFCD11044971
CAS Number: 959755-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR