2-Fluoro-4-nitropyridine ≥97%

Supplier: Ambeed
Danger

A774166-5G A774166-100MG A774166-1G A774166-250MG
77614-370EA 763.23 USD
77614-370 77891-517 77891-518 77891-519
2-Fluoro-4-nitropyridine ≥97%
2-Fluoro-4-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃FN₂O₂
MDL Number: MFCD04112489
CAS Number: 18614-46-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR