2-Amino-4,4,4-trifluorobutyric acid hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-2-Amino-4,4,4-trifluorobutyric acid hydrochloride
A773563-250MG
A773563-1G
77614-368EA
88.58
USD
77614-368
77674-124
2-Amino-4,4,4-trifluorobutyric acid hydrochloride ≥97%
2-Amino-4,4,4-trifluorobutyric acid hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₇ClF₃NO₂ |
MDL Number:
MFCD00671488 CAS Number: 15959-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...