4-(4-Morpholinyl)phenol ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-Morpholinophenol

A773456-100MG A773456-250MG
77749-529EA 23 USD
77749-529 77749-530
4-(4-Morpholinyl)phenol ≥97%
4-(4-Morpholinyl)phenol

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.2157 g/mol
MDL Number: MFCD00066933
CAS Number: 6291-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR