3-Methoxy-4-(4-methylpiperazin-1-yl)aniline ≥98%

Supplier: Ambeed
Danger

A772757-1G A772757-250MG
77674-120EA 292 USD
77674-120 77674-122
3-Methoxy-4-(4-methylpiperazin-1-yl)aniline ≥98%
3-Methoxy-4-(4-methylpiperazin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O
MW: 221.3 g/mol
MDL Number: MFCD09909622
CAS Number: 156428-85-2
UN: 3263
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR