CUDC-305 ≥98%

Supplier: Ambeed
Warning

Synonyms: Debio 0932, 2-((6-(Dimethylamino)benzo[d][1,3]dioxol-5-yl)thio)-1-(2-(neopentylamino)ethyl)-1H-imidazo[4,5-c]pyridin-4-amine

A772543-2MG A772543-10MG A772543-5MG
77749-527EA 82 USD
77749-527 77749-526 77749-528
CUDC-305 ≥98%
CUDC-305

Precaución: for Research Use Only

Formula: C₂₂H₃₀N₆O₂S
MW: 442.58 g/mol
MDL Number: MFCD18633209
CAS Number: 1061318-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR