Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-O-((2S,3R,4R,5R,6R)-3-acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)-L-threonine, Fmoc-Thr[GalNAc(Ac)3-α-D]-OH, Fmoc-L-Thr(α-D-GalNAc(Ac)3)-OH

A772064-100MG A772064-50MG
77670-066EA 463 USD
77670-066 77880-549
Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH ≥98%
Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH

Precaución: for Research Use Only

Formula: C₃₃H₃₈N₂O₁₃
MDL Number: MFCD00171374
CAS Number: 116783-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR