1-Methylindole-2-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A771353-250MG A771353-5G
77884-707EA 31 USD
77884-707 77884-708
1-Methylindole-2-boronic acid pinacol ester ≥98%
1-Methylindole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₂
MW: 257.135 g/mol
MDL Number: MFCD13182063
CAS Number: 596819-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR