4-(Methylamino)butanoic acid ≥98%

Supplier: Ambeed
Warning

A770024-100MG A770024-1G A770024-250MG
77595-218EA 13.33 USD
77595-218 77670-060 77670-062
4-(Methylamino)butanoic acid ≥98%
4-(Methylamino)butanoic acid

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂
MW: 117.14 g/mol
MDL Number: MFCD00044627
CAS Number: 1119-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR