Ethylene glycol mono(4-bromophenyl)ether ≥98%

Supplier: Ambeed
Warning

A769679-1G A769679-25G A769679-100G A769679-5G
77627-534EA 17.51 USD
77627-534 77627-536 77878-615 77878-616
Ethylene glycol mono(4-bromophenyl)ether ≥98%
Ethylene glycol mono(4-bromophenyl)ether

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MDL Number: MFCD00002860
CAS Number: 34743-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR