Fmoc-L-(2,6-di-Me)Tyr-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Tyr(2,6-DiMe)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxy-2,6-dimethylphenyl)propanoic acid, 2,6-Dimethyl-L-tyrosine, N-FMOC protected

A769111-1G A769111-250MG
77573-478EA 83.43 USD
77573-478 77684-710
Fmoc-L-(2,6-di-Me)Tyr-OH ≥98%
Fmoc-L-(2,6-di-Me)Tyr-OH

Precaución: For research use only.

Formula: C₂₆H₂₅NO₅
MW: 431.49 g/mol
MDL Number: MFCD01632251
CAS Number: 206060-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR