3-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid ≥90%

Supplier: Ambeed
Warning

A768882-100MG A768882-250MG
77893-519EA 97 USD
77893-519 77893-520
3-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid ≥90%
3-(1-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD12913674
CAS Number: 165949-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR