2-Chloro-3H-quinazolin-4-one ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloroquinazolin-4(3H)-one

A768378-250MG A768378-1G A768378-5G
77674-112EA 30 USD
77674-112 77884-704 77884-705
2-Chloro-3H-quinazolin-4-one ≥97%
2-Chloro-3H-quinazolin-4-one

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MDL Number: MFCD04086742
CAS Number: 607-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR