4,7-Diazaspiro[2.5]octane-5,8-dione ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A768059-1G A768059-5G A768059-10G
77684-706EA 128 USD
77684-706 77891-504 77891-503
4,7-Diazaspiro[2.5]octane-5,8-dione ≥95%, Ambeed.Inc
4,7-Diazaspiro[2.5]octane-5,8-dione

Precaución: For research use only.

Formula: C₆H₈N₂O₂
MDL Number: MFCD18249952
CAS Number: 129306-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR