Ethyl 2-(5-aminopyridin-2-yl)acetate ≥97%

Supplier: Ambeed
Warning

A768007-100MG A768007-1G A768007-250MG
77886-734EA 54 USD
77886-734 77886-735 77886-736
Ethyl 2-(5-aminopyridin-2-yl)acetate ≥97%
Ethyl 2-(5-aminopyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD20696931
CAS Number: 174890-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR