4,5,6,7-Tetrabromo-N,N-dimethyl-1H-benzo[d]imidazol-2-amine ≥98%
Supplier: Ambeed
|
Warning
|
A767723-100MG
A767723-10MG
A767723-1G
A767723-5MG
A767723-250MG
A767723-50MG
77686-779EA
34
USD
77686-779
77686-780
77686-781
77686-782
77901-575
77901-483
4,5,6,7-Tetrabromo-N,N-dimethyl-1H-benzo[d]imidazol-2-amine ≥98%
4,5,6,7-Tetrabromo-N,N-dimethyl-1H-benzo[d]imidazol-2-amine
Precaución: for Research Use Only
Formula:
C₉H₇Br₄N₃ |
MDL Number:
MFCD08705322 CAS Number: 749234-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...