Ammonium benzoate ≥99%

Supplier: Ambeed
Warning

A767496-100G A767496-1000G A767496-500G
77627-520EA 5.15 USD
77627-520 77878-610 77878-611
Ammonium benzoate ≥99%
Ammonium benzoate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD00013069
CAS Number: 1863-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR