2,5-Dioxopyrrolidin-1-yl 4-(pyridin-2-yldisulfaneyl)pentanoate ≥98%

Supplier: Ambeed
Warning

A767440-1G A767440-250MG
77573-474EA 387 USD
77573-474 77686-778
2,5-Dioxopyrrolidin-1-yl 4-(pyridin-2-yldisulfaneyl)pentanoate ≥98%
2,5-Dioxopyrrolidin-1-yl 4-(pyridin-2-yldisulfaneyl)pentanoate

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₄S₂
MDL Number: MFCD20230233
CAS Number: 341498-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR