2-(3-(3-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)benzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)-3-ethylbenzo[d]oxazol-3-ium iodide ≥98%

Supplier: Ambeed
Warning

Synonyms: Cy2-SE (iodine)

A767344-100MG A767344-50MG A767344-250MG A767344-1MG
77747-516EA 531 USD
77747-516 77747-519 77747-518 77747-517
2-(3-(3-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)benzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)-3-ethylbenzo[d]oxazol-3-ium iodide ≥98%
2-(3-(3-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)benzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)-3-ethylbenzo[d]oxazol-3-ium iodide

Precaución: for Research Use Only

Formula: C₂₉H₃₀IN₃O₆
MW: 643.46 g/mol
MDL Number: MFCD29472246
CAS Number: 186205-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR